Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000326194.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000355092.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000355961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000359882.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000361988.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000367574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000472520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000485288.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000491243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000537758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_001282167.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_022825.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_203473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_203474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_203475.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
TC0X00007190.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000359882.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000326194.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000355092.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000355961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000361988.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000367574.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000472520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000485288.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000491243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
ENST00000537758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_001282167.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_022825.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_203473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_203474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149048.hg.1 |
NM_203475.2 |
Refseq |