Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000609076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000609076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000609076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000609076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000609494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000609494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000609494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
ENST00000609494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004633.hg.1 |
ENST00000262811.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004633.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004633.hg.1 |
TC19000351.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004634.hg.1 |
ENST00000262811.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004634.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19004634.hg.1 |
TC19000351.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
TC1900007363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153933.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000609076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000609076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000609076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000609076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000609494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000609494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000609494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
ENST00000609494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007363.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
NM_015016.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
TC1900007363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
ENST00000262811.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
ENST00000601226.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
ENST00000608648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900153934.hg.1 |
NM_015016.1 |
Refseq |