Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01110394_m1
          See other ITGB8 GE Assays ›
          Gene Symbol:
          ITGB8
          Gene Name:
          integrin subunit beta 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 20329819 - 20415759 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01110394_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITGB8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002214.2 NP_002205.1 5 - 6 1511 64
          XM_011515393.2 XP_011513695.1 6 - 7 1314 64
          XM_011515394.2 XP_011513696.1 6 - 7 912 64
          XM_011515396.1 XP_011513698.1 6 - 7 1583 64
          XM_017012178.1 XP_016867667.1 6 - 7 1208 64
          XM_017012179.1 XP_016867668.1 6 - 7 1667 64
          XM_017012180.1 XP_016867669.1 6 - 7 1960 64
          XM_017012181.1 XP_016867670.1 7 - 8 1779 64
          XM_017012182.1 XP_016867671.1 6 - 7 894 64
          XM_017012183.1 XP_016867672.1 6 - 7 851 64
          GenBank mRNA AK295044.1 - 6 - 7 1583 64
          BC002630.2 - 5 - 6 1511 3609490 64
          BT006739.1 - 5 - 6 806 64
          M73780.1 - 5 - 6 1486 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700006844.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000125.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human Phagocytosis of Microbes 1700 Inventoried Human_Phagocytosis_of_Microbes Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITGB8

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          ganglioside metabolic process
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          positive regulation of gene expression
          negative regulation of gene expression
          extracellular matrix organization
          cartilage development
          placenta blood vessel development
          receptor activity
          receptor binding
          extracellular matrix protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07001915.hg.1 TC07000125.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07001915.hg.1 ENST00000222573.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07001915.hg.1 ENST00000537992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07001915.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000460204.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000460204.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700147293.hg.1 TC0700006844.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000222573.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000460204.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000460204.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000477859.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000478974.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 ENST00000537992.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Affymetrix Human Clariom D Assay TC0700006844.hg.1 NM_002214.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 230944 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0