Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01111241_m1
          See other ZNF271P GE Assays ›
          Gene Symbol:
          ZNF271P
          Gene Name:
          zinc finger protein 271, pseudogene
          Gene Aliases:
          CT-ZFP48, HZF7, ZNF-EB, ZNF-dp, ZNF271, ZNFEB
          Chromosome Location:
          Chr.18: 35290272 - 35310766 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01111241_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNF271P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024565.1 1 - 2 117 98
          GenBank mRNA AK023456.1 - 1 - 2 56 98
          DA005750.1 - 1 - 2 84 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA575740.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306059 [+] on Build GRCh38 98
          GenBankmRNA HY167884.1 - 1 - 2 73 Chr. 18 - 35290316 - 35306154 [+] on Build GRCh38 98
          GenBankmRNA DA683872.1 - 1 - 2 73 Chr. 18 - 35290316 - 35306168 [+] on Build GRCh38 98
          GenBankmRNA DA479011.1 - 1 - 2 84 Chr. 18 - 35290305 - 35306159 [+] on Build GRCh38 98
          GenBankmRNA DA703328.1 - 1 - 2 56 Chr. 18 - 35290333 - 35306103 [+] on Build GRCh38 98
          GenBankmRNA HY159347.1 - 1 - 2 46 Chr. 18 - 35290345 - 35306192 [+] on Build GRCh38 98
          GenBankmRNA DB201677.1 - 1 - 2 73 Chr. 18 - 35290316 - 35306274 [+] on Build GRCh38 98
          GenBankmRNA DB209506.1 - 1 - 2 73 Chr. 18 - 35290316 - 35306210 [+] on Build GRCh38 98
          GenBankmRNA DA954621.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306219 [+] on Build GRCh38 98
          GenBankmRNA DA347187.1 - 1 - 2 56 Chr. 18 - 35290333 - 35306260 [+] on Build GRCh38 98
          GenBankmRNA DB206403.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306269 [+] on Build GRCh38 98
          GenBankmRNA BF212348.1 - 108 Chr. - - [-] on Build 98
          GenBankmRNA DA720988.1 - 1 - 2 51 Chr. 18 - 35290338 - 35306260 [+] on Build GRCh38 98
          GenBankmRNA DA517516.1 - 1 - 2 73 Chr. 18 - 35290316 - 35306211 [+] on Build GRCh38 98
          GenBankmRNA DB048870.1 - 1 - 2 66 Chr. 18 - 35290324 - 35306252 [+] on Build GRCh38 98
          GenBankmRNA DA965582.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306258 [+] on Build GRCh38 98
          GenBankmRNA DB201350.1 - 1 - 2 66 Chr. 18 - 35290323 - 35306188 [+] on Build GRCh38 98
          GenBankmRNA DA733689.1 - 1 - 2 82 Chr. 18 - 35290307 - 35306279 [+] on Build GRCh38 98
          GenBankmRNA DA375478.1 - 1 - 2 56 Chr. 18 - 35290333 - 35306241 [+] on Build GRCh38 98
          GenBankmRNA BU587637.1 - 1 - 2 68 Chr. 18 - 35290324 - 35306164 [+] on Build GRCh38 98
          GenBankmRNA DA692276.1 - 1 - 2 67 Chr. 18 - 35290322 - 35306258 [+] on Build GRCh38 98
          GenBankmRNA DA573545.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306216 [+] on Build GRCh38 98
          GenBankmRNA DB052152.1 - 1 - 2 73 Chr. 18 - 35290316 - 35306245 [+] on Build GRCh38 98
          GenBankmRNA DA133574.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306216 [+] on Build GRCh38 98
          GenBankmRNA DA576313.1 - 1 - 2 82 Chr. 18 - 35290307 - 35306223 [+] on Build GRCh38 98
          GenBankmRNA DB121554.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306216 [+] on Build GRCh38 98
          GenBankmRNA DA002482.1 - 1 - 2 65 Chr. 18 - 35290325 - 35306192 [+] on Build GRCh38 98
          GenBankmRNA BI824898.1 - Chr. - - [-] on Build 99
          GenBankmRNA DA811982.1 - 1 - 2 74 Chr. 18 - 35290324 - 35306324 [+] on Build GRCh38 98
          GenBankmRNA DA965096.1 - 1 - 2 56 Chr. 18 - 35290333 - 35306191 [+] on Build GRCh38 98
          GenBankmRNA DA877937.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306256 [+] on Build GRCh38 98
          GenBankmRNA CK003080.1 - 1 - 2 60 Chr. 18 - 35290329 - 35306310 [+] on Build GRCh38 98
          GenBankmRNA DA308659.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306250 [+] on Build GRCh38 98
          GenBankmRNA DA831167.1 - 1 - 2 75 Chr. 18 - 35290314 - 35306419 [+] on Build GRCh38 98
          GenBankmRNA DA748162.1 - 1 - 2 75 Chr. 18 - 35290314 - 35306241 [+] on Build GRCh38 98
          GenBankmRNA DA735321.1 - 1 - 2 84 Chr. 18 - 35290305 - 35306430 [+] on Build GRCh38 98
          GenBankmRNA AU135134.1 - 1 - 2 56 Chr. 18 - 35290333 - 35306344 [+] on Build GRCh38 98
          GenBankmRNA DB128388.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306258 [+] on Build GRCh38 98
          GenBankmRNA BP261844.1 - 1 - 2 65 Chr. 18 - 35290324 - 35306266 [+] on Build GRCh38 98
          GenBankmRNA BU935462.1 - 1 - 2 57 Chr. 18 - 35290334 - 35306030 [+] on Build GRCh38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger and SCAN domain containing 30 ZSCAN30 ZNF-WYM,
          ZNF397OS,
          ZNF917
          -65.0 Anti-sense Chr. 18 - 35251058 - 35290261 on NCBI Build GRCh38
          Downstream zinc finger protein 24 ZNF24 KOX17,
          RSG-A,
          ZNF191,
          ZSCAN3,
          Zfp191
          41.0kb Anti-sense Chr. 18 - 35332208 - 35344467 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800009224.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000144.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK023456.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR18001631.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18001631.hg.1 NR_024565.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18001631.hg.1 NR_024566.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18001631.hg.1 TC18000144.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18001634.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18001634.hg.1 NR_024565.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18001634.hg.1 TC18000144.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 ENST00000465539.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 ENST00000465539.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024566.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024566.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024566.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800009224.hg.1 NR_024566.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 ENST00000465539.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 ENST00000465539.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024566.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024566.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024566.1 Refseq
          Affymetrix Human Clariom D Assay TC1800009224.hg.1 NR_024566.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800163488.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800163488.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800163488.hg.1 NR_024566.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800163488.hg.1 TC1800009224.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800163488.hg.1 NR_024566.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800163488.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800163488.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800163489.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800163489.hg.1 NR_024565.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800163489.hg.1 TC1800009224.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800163489.hg.1 ENST00000399070.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1800163489.hg.1 NR_024565.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0