Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01112297_g1
          See other PPP1R8 GE Assays ›
          Gene Symbol:
          PPP1R8
          Gene Name:
          protein phosphatase 1 regulatory subunit 8
          Gene Aliases:
          ARD-1, ARD1, NIPP-1, NIPP1, PRO2047
          Chromosome Location:
          Chr.1: 27830741 - 27851672 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01112297_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPP1R8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014110.4 NP_054829.2 4 - 5 584 81
          NM_138558.2 NP_612568.1 4 - 5 866 81
          GenBank mRNA AF061958.1 - 4 - 5 512 81
          AF061959.1 - 4 - 5 667 81
          AF126488.1 - 4 - 5 500 81
          AK292077.1 - 4 - 5 512 81
          AK304619.1 - 4 - 5 512 81
          AX775985.1 - 4 - 5 500 81
          BC001597.1 - 3 - 4 392 81
          BC013360.2 - 3 - 4 442 3633676 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100007538.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PPP1R8

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          RNA catabolic process
          cell population proliferation
          RNA splicing
          negative regulation of catalytic activity
          RNA phosphodiester bond hydrolysis
          DNA binding
          RNA binding
          endonuclease activity
          protein serine/threonine phosphatase inhibitor activity
          protein binding
          ribonuclease E activity
          protein phosphatase regulator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000486634.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 ENST00000486634.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 TC0100007538.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100152717.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_014110.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_138558.2 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000236412.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000311772.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000373931.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000431586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000486634.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 ENST00000486634.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007538.hg.1 NM_002713.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0