Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01113531_m1
          See other CDC73 GE Assays ›
          Gene Symbol:
          CDC73
          Gene Name:
          cell division cycle 73
          Gene Aliases:
          C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
          Chromosome Location:
          Chr.1: 193121958 - 193254815 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01113531_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDC73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024529.4 NP_078805.3 16 - 17 1797 97
          GenBank mRNA AF312865.1 - 16 - 17 1562 97
          AK026969.1 - 16 - 17 1687 97
          AK226038.1 - 16 - 17 1694 97
          BC014351.1 - 12 - 13 1188 97
          BC065037.1 - 16 - 17 1634 6170851 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100011027.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001626.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDC73

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          endodermal cell fate commitment
          transcription elongation from RNA polymerase II promoter
          mRNA polyadenylation
          cell cycle
          negative regulation of cell population proliferation
          histone monoubiquitination
          Wnt signaling pathway
          stem cell population maintenance
          positive regulation of Wnt signaling pathway
          positive regulation of mRNA 3'-end processing
          protein destabilization
          positive regulation of transcription elongation from RNA polymerase II promoter
          histone H2B ubiquitination
          negative regulation of myeloid cell differentiation
          positive regulation of transcription by RNA polymerase II
          negative regulation of fibroblast proliferation
          negative regulation of epithelial cell proliferation
          cellular response to lipopolysaccharide
          beta-catenin-TCF complex assembly
          negative regulation of G1/S transition of mitotic cell cycle
          RNA polymerase II complex binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01025152.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025152.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01025152.hg.1 TC01001626.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01025153.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01025153.hg.1 TC01001626.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01025150.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025150.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025150.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01025150.hg.1 TC01001626.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01025151.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01025151.hg.1 NM_024529.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01025151.hg.1 TC01001626.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 ENST00000482484.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay TC0100011027.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180946.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180946.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180946.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180946.hg.1 TC0100011027.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180946.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180946.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180946.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180949.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180949.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180949.hg.1 TC0100011027.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180949.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180949.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180948.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180948.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180948.hg.1 TC0100011027.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180948.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180948.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180947.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180947.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180947.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180947.hg.1 TC0100011027.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180947.hg.1 ENST00000367435.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180947.hg.1 ENST00000477868.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100180947.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180950.hg.1 NM_024529.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100180950.hg.1 TC0100011027.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100180950.hg.1 NM_024529.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline