Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004446.2 | NP_004437.2 | 24 - 25 | 3723 | 62 | |
XM_017000614.1 | XP_016856103.1 | 13 - 14 | 1979 | 62 | ||
GenBank mRNA | AK298192.1 | - | 5 - 6 | 547 | 62 | |
AY493416.1 | - | 22 - 23 | 3270 | 62 | ||
BC126275.1 | - | 24 - 25 | 3591 | 8991991 | 62 | |
BC136465.1 | - | 24 - 25 | 3491 | 9020454 | 62 | |
CR933648.1 | - | 24 - 25 | 3563 | 62 | ||
X54326.1 | - | 22 - 23 | 3295 | 62 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |