Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000549864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000549864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
ENST00000549864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
NR_110019.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
NR_110019.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
NR_110019.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
NR_110020.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
NR_110020.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007530.hg.1 |
NR_110020.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200152902.hg.1 |
ENST00000549864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200152902.hg.1 |
NR_110020.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200152902.hg.1 |
TC1200007530.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200152902.hg.1 |
ENST00000549864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200152902.hg.1 |
NR_110020.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000547774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000549864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000549864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
ENST00000549864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
NR_110019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
NR_110019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
NR_110019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
NR_110020.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
NR_110020.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007530.hg.1 |
NR_110020.2 |
Refseq |