Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01115099_g1
          See other LOC100506125 GE Assays ›
          Gene Symbol:
          LOC100506125
          Gene Name:
          uncharacterized LOC100506125
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 48789147 - 48790535 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01115099_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100506125
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110019.1 2 - 3 216 100
          NR_110020.1 2 - 3 250 100
          GenBank mRNA BX098326.1 - 2 - 3 216 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI675757.1 - 1 - 2 Chr. 12 - 48789576 - 48790514 [-] on Build GRCh38 100
          GenBankmRNA HY332360.1 - 1 - 2 Chr. 12 - 48789610 - 48790528 [-] on Build GRCh38 100
          GenBankmRNA BM758062.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790535 [+] on Build GRCh38 100
          GenBankmRNA BM757607.1 - 2 - 3 168 Chr. 12 - 48789661 - 48790467 [+] on Build GRCh38 100
          GenBankmRNA AI458332.1 - 1 - 2 Chr. 12 - 48789183 - 48790528 [-] on Build GRCh38 100
          GenBankmRNA BM757504.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790537 [+] on Build GRCh38 100
          GenBankmRNA BM758623.1 - 2 - 3 167 Chr. 12 - 48789661 - 48790535 [+] on Build GRCh38 100
          GenBankmRNA BM748284.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790537 [+] on Build GRCh38 100
          GenBankmRNA BM758444.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790535 [+] on Build GRCh38 100
          GenBankmRNA BF195775.1 - 1 - 2 Chr. 12 - 48790012 - 48790549 [-] on Build GRCh38 100
          GenBankmRNA BM748483.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790537 [+] on Build GRCh38 100
          GenBankmRNA BM758004.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790534 [+] on Build GRCh38 100
          GenBankmRNA BM748296.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790522 [+] on Build GRCh38 100
          GenBankmRNA AI391488.1 - 1 - 2 Chr. 12 - 48789180 - 48790537 [-] on Build GRCh38 100
          GenBankmRNA BM749854.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790537 [+] on Build GRCh38 100
          GenBankmRNA BM758340.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790530 [+] on Build GRCh38 100
          GenBankmRNA HY191907.1 - 1 - 2 Chr. 12 - 48789611 - 48790555 [-] on Build GRCh38 100
          GenBankmRNA BM757608.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790535 [+] on Build GRCh38 100
          GenBankmRNA AI436693.1 - 1 - 2 Chr. 12 - 48789983 - 48790548 [-] on Build GRCh38 100
          GenBankmRNA AI686141.1 - 1 - 2 Chr. 12 - 48789180 - 48790535 [-] on Build GRCh38 100
          GenBankmRNA HY007981.1 - 2 - 3 305 Chr. 12 - 48789522 - 48790559 [+] on Build GRCh38 100
          GenBankmRNA BM743364.1 - 2 - 3 165 Chr. 12 - 48789661 - 48790537 [+] on Build GRCh38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream adenylate cyclase 6 ADCY6 AC6,
          LCCS8
          -948.0 Anti-sense Chr. 12 - 48766191 - 48789096 on NCBI Build GRCh38
          Downstream calcium voltage-gated channel auxiliary subunit beta 3 CACNB3 CAB3,
          CACNLB3
          23.9kb Sense Chr. 12 - 48814432 - 48828943 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0