Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01115662_m1
          See other TNC GE Assays ›
          Gene Symbol:
          TNC
          Gene Name:
          tenascin C
          Gene Aliases:
          150-225, DFNA56, GMEM, GP, HXB, JI, TN, TN-C
          Chromosome Location:
          Chr.9: 115019575 - 115118257 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01115662_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNC
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002160.3 NP_002151.2 25 - 26 6584 78
          XM_005251972.3 XP_005252029.1 24 - 25 6261 78
          XM_005251973.3 XP_005252030.1 21 - 22 5442 78
          XM_005251974.3 XP_005252031.1 19 - 20 4896 78
          XM_005251975.3 XP_005252032.1 18 - 19 4623 78
          XM_006717096.3 XP_006717159.1 26 - 27 6810 78
          XM_006717097.3 XP_006717160.1 24 - 25 6261 78
          XM_006717098.3 XP_006717161.1 23 - 24 5988 78
          XM_006717101.3 XP_006717164.1 19 - 20 4896 78
          XM_011518625.2 XP_011516927.1 23 - 24 5988 78
          XM_011518626.2 XP_011516928.1 22 - 23 5718 78
          XM_011518628.2 XP_011516930.1 20 - 21 5169 78
          XM_011518629.2 XP_011516931.1 20 - 21 5169 78
          XM_017014678.1 XP_016870167.1 27 - 28 7083 78
          XM_017014679.1 XP_016870168.1 26 - 27 6810 78
          XM_017014680.1 XP_016870169.1 26 - 27 6807 78
          XM_017014681.1 XP_016870170.1 23 - 24 5991 78
          GenBank mRNA AB210038.1 - 25 - 26 6460 78
          BC151843.1 - 22 - 23 5633 8860362 78
          GU473235.1 - 20 - 21 84
          M55618.1 - 24 - 25 84
          X56160.1 - 24 - 25 6223 78
          X78565.1 - 25 - 26 6485 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011305.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001517.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNC

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          cell adhesion
          negative regulation of cell adhesion
          neuromuscular junction development
          positive regulation of cell population proliferation
          response to wounding
          response to mechanical stimulus
          positive regulation of gene expression
          peripheral nervous system axon regeneration
          extracellular matrix organization
          wound healing
          odontogenesis of dentin-containing tooth
          response to ethanol
          bud outgrowth involved in lung branching
          mesenchymal-epithelial cell signaling involved in prostate gland development
          prostate gland epithelium morphogenesis
          cellular response to retinoic acid
          cellular response to vitamin D
          response to fibroblast growth factor
          cellular response to prostaglandin D stimulus
          syndecan binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000340094.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000341037.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000345230.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000346706.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000350763.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000423613.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000535648.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000537320.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 ENST00000542877.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 NM_002160.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09019001.hg.1 TC09001517.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000341037.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000350763.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000423613.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000535648.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000537320.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000542877.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 NM_002160.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 TC0900011305.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000341037.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000350763.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000423613.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000535648.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000537320.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 ENST00000542877.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900177106.hg.1 NM_002160.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0