Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000514928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000514928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
ENST00000514928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022690.hg.1 |
ENST00000225969.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022690.hg.1 |
ENST00000442592.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022690.hg.1 |
ENST00000503633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022690.hg.1 |
ENST00000507088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022690.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022690.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022690.hg.1 |
TC17001685.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
TC1700011096.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189371.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000225969.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000442592.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000503633.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000507088.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000508200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000511860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000514928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000514928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
ENST00000514928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011096.hg.1 |
NM_016504.2 |
Refseq |