Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008530.2 | NP_001008530.1 | 6 - 7 | 764 | 79 | |
NM_005606.6 | NP_005597.3 | 5 - 6 | 646 | 79 | ||
XM_005267862.3 | XP_005267919.1 | 5 - 6 | 632 | 79 | ||
XM_005267863.3 | XP_005267920.1 | 5 - 6 | 632 | 79 | ||
XM_011536970.1 | XP_011535272.1 | 5 - 6 | 632 | 79 | ||
XM_017021463.1 | XP_016876952.1 | 5 - 6 | 456 | 79 | ||
XM_017021464.1 | XP_016876953.1 | 5 - 6 | 561 | 79 | ||
XM_017021465.1 | XP_016876954.1 | 5 - 6 | 450 | 79 | ||
XM_017021466.1 | XP_016876955.1 | 5 - 6 | 539 | 79 | ||
XM_017021467.1 | XP_016876956.1 | 5 - 6 | 550 | 79 | ||
GenBank mRNA | AK225822.1 | - | 5 - 6 | 537 | 79 | |
AK291534.1 | - | 5 - 6 | 571 | 79 | ||
AK291560.1 | - | 5 - 6 | 571 | 79 | ||
AK297074.1 | - | 5 - 6 | 466 | 79 | ||
AK297780.1 | - | 6 - 7 | 503 | 79 | ||
BC003061.2 | - | 5 - 6 | 535 | 3504506 | 79 | |
BX161380.1 | - | 5 - 6 | 552 | 79 | ||
BX161422.1 | - | 5 - 6 | 549 | 79 | ||
BX161434.1 | - | 6 - 7 | 674 | 79 | ||
CR457406.1 | - | 4 - 5 | 403 | 79 | ||
Y09862.1 | - | 5 - 6 | 473 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |