Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01120011_s1
          See other RHPN1-AS1 GE Assays ›
          Gene Symbol:
          RHPN1-AS1
          Gene Name:
          RHPN1 antisense RNA 1 (head to head)
          Gene Aliases:
          C8orf51
          Chromosome Location:
          Chr.8: 143366623 - 143368635 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01120011_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RHPN1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026785.1 1 1885 96
          GenBank mRNA AK026503.1 - 1 1795 96
          BC000203.1 - 1 1798 96
          BM685262.1 - 1 96
          BM968345.1 - 1 463 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW139464.1 - 1 Chr. 8 - 143366636 - 143366991 [+] on Build GRCh38 96
          GenBankmRNA AW139092.1 - 1 Chr. 8 - 143366631 - 143366898 [+] on Build GRCh38 96
          GenBankmRNA BM684166.1 - 1 Chr. 8 - 143366621 - 143367218 [+] on Build GRCh38 96
          GenBankmRNA BM930735.1 - 1 500 Chr. 8 - 143366621 - 143367250 [-] on Build GRCh38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream topoisomerase (DNA) I,
          mitochondrial
          TOP1MT -6.7kb Anti-sense Chr. 8 - 143309324 - 143359977 on NCBI Build GRCh38
          Downstream rhophilin Rho GTPase binding protein 1 RHPN1 ODF5,
          RHOPHILIN,
          RHPN
          1.9kb Sense Chr. 8 - 143368827 - 143384220 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 195380 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 273456 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P71503 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P71503 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 40286 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42024 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P16709593 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P71503 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P71503 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16146286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817019 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532296 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532289 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16146287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16146282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532288 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532291 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532322 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532308 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532311 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532320 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532291 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16146282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532308 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817019 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817032 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532311 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817014 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16146287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16146286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817016 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817030 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532320 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532296 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532322 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26817039 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532288 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06532289 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P71503 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P122700 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P71503 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 1483 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 3567 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16709593 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32363438 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532305 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16146285 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532288 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532282 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532311 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532290 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532293 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16146277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16146288 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532306 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532334 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532285 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532298 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532330 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532308 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16146287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532332 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532281 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532275 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532325 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532300 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16146282 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532279 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532327 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532307 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16146274 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532323 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532335 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06532296 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P16709593 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8153406 - -
          Affymetrix Human Gene 1.0 ST Array 8153408 - -
          Affymetrix Human Gene 1.0 ST Array 8153407 - -
          Affymetrix Human Gene 1.1 ST Array 8153405 - -
          Affymetrix HT HG-U133+ PM Array Plate 207458_PM_at - -
          Human Genome U133 Plus 2.0 Array 207458_PM_at - -
          Human Genome U133 Plus 2.0 Array 207458_at - -
          Human Genome U133A 2.0 Array 207458_PM_at - -
          Human Genome U133A 2.0 Array 207458_at - -
          AffymetrixPathwork Diagnostics Pathchip pw597873 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207458_at4 - -
          Human Genome U133 Set 207458_at - -
          Human Genome U133 Set 207458_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036214 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036214 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 137078 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1688772 BM685262.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1688772 BC000203.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1688772 BM968345.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1688772 NR_026785.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1688772 AK026503.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1688772 BM685262.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1688772 BC000203.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1688772 BM968345.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1688772 NR_026785.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1688772 AK026503.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1688772 BM685262.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1688772 BC000203.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1688772 BM968345.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1688772 NR_026785.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1688772 AK026503.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1688772 BM685262.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1688772 BC000203.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1688772 BM968345.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1688772 NR_026785.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1688772 AK026503.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1688772 BM685262.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1688772 BC000203.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1688772 BM968345.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1688772 NR_026785.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1688772 AK026503.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1688772 BM685262.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1688772 BC000203.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1688772 BM968345.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1688772 NR_026785.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1688772 AK026503.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0