Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01121937_m1
See other ARHGEF11 GE Assays ›
Gene Symbol:
ARHGEF11
Gene Name:
Rho guanine nucleotide exchange factor 11
Gene Aliases:
GTRAP48, PDZ-RHOGEF
Chromosome Location:
Chr.1: 156934840 - 157046903 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01121937_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ARHGEF11
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_014784.3
NP_055599.1
16 - 17
2421
52
NM_198236.2
NP_937879.1
17 - 18
2541
52
XM_005245629.4
XP_005245686.1
17 - 18
2942
52
XM_005245633.4
XP_005245690.1
7 - 8
804
52
XM_006711659.2
XP_006711722.1
19 - 20
2990
52
XM_006711660.3
XP_006711723.1
19 - 20
2990
52
XM_006711661.2
XP_006711724.1
19 - 20
2990
52
XM_006711662.2
XP_006711725.1
19 - 20
2990
52
XM_006711663.3
XP_006711726.1
17 - 18
1977
52
XM_006711665.3
XP_006711728.1
18 - 19
2870
52
XM_011510185.2
XP_011508487.1
18 - 19
2975
52
XM_011510186.2
XP_011508488.1
18 - 19
2870
52
XM_011510187.2
XP_011508489.1
18 - 19
1701
52
XM_017002920.1
XP_016858409.1
18 - 19
2957
52
XM_017002921.1
XP_016858410.1
17 - 18
2942
52
XM_017002922.1
XP_016858411.1
18 - 19
2870
52
XM_017002923.1
XP_016858412.1
16 - 17
2822
52
XM_017002924.1
XP_016858413.1
17 - 18
2855
52
GenBank mRNA
AB002378.1
-
16 - 17
2126
52
BC057394.1
-
17 - 18
2565
6148684
52
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ARHGEF11
Gene Ontology Categories:
Function(s)
Process(es)
cytokinesis
regulation of cell growth
movement of cell or subcellular component
striated muscle contraction
G protein-coupled receptor signaling pathway
Rho protein signal transduction
establishment of cell polarity
actin cytoskeleton organization
regulation of Rho protein signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
positive regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
G protein-coupled receptor binding
guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0