Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01122276_m1
          See other NIPBL GE Assays ›
          Gene Symbol:
          NIPBL
          Gene Name:
          NIPBL, cohesin loading factor
          Gene Aliases:
          CDLS, CDLS1, IDN3, IDN3-B, Scc2
          Chromosome Location:
          Chr.5: 36876759 - 37065819 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01122276_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NIPBL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015384.4 NP_056199.2 42 - 43 7758 107
          NM_133433.3 NP_597677.2 42 - 43 7758 107
          XM_005248280.3 XP_005248337.1 42 - 43 7747 107
          XM_005248282.4 XP_005248339.3 36 - 37 7249 107
          XM_006714468.2 XP_006714531.1 41 - 42 7549 107
          XM_017009330.1 XP_016864819.1 34 - 35 10021 107
          XM_017009331.1 XP_016864820.1 41 - 42 6121 107
          GenBank mRNA AJ627032.1 - 42 - 43 7727 107
          AJ640137.1 - 42 - 43 7727 107
          AK002065.1 - 6 - 7 876 107
          AK022659.1 - 8 - 9 1070 107
          AK023416.1 - 10 - 11 1406 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500007163.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000166.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NIPBL

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          metanephros development
          heart morphogenesis
          outflow tract morphogenesis
          cellular response to DNA damage stimulus
          mitotic sister chromatid cohesion
          brain development
          sensory perception of sound
          stem cell population maintenance
          positive regulation of histone deacetylation
          maintenance of mitotic sister chromatid cohesion
          cellular protein localization
          embryonic forelimb morphogenesis
          forelimb morphogenesis
          external genitalia morphogenesis
          positive regulation of multicellular organism growth
          ear morphogenesis
          regulation of hair cycle
          fat cell differentiation
          positive regulation of ossification
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of embryonic development
          embryonic digestive tract morphogenesis
          developmental growth
          eye morphogenesis
          regulation of developmental growth
          embryonic viscerocranium morphogenesis
          cognition
          face morphogenesis
          gall bladder development
          uterus morphogenesis
          cellular response to X-ray
          chromatin binding
          protein binding
          protein C-terminus binding
          mediator complex binding
          histone deacetylase binding
          protein N-terminus binding
          chromo shadow domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05001946.hg.1 ENST00000282516.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05001946.hg.1 ENST00000448238.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05001946.hg.1 NM_015384.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05001946.hg.1 NM_133433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05001946.hg.1 TC05000166.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 ENST00000282516.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 ENST00000448238.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 ENST00000514335.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 NM_015384.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 NM_133433.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 TC0500007163.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 NM_015384.4 Refseq
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 ENST00000282516.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 ENST00000448238.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 ENST00000514335.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500148511.hg.1 NM_133433.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0