Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008624.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008624.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008624.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008624.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008624.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008624.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160396.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160396.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160396.hg.1 |
TC2000008624.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160396.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160396.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008624.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008624.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008624.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008624.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008624.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008624.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160394.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160394.hg.1 |
NR_110615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160394.hg.1 |
TC2000008624.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160394.hg.1 |
ENST00000420928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160394.hg.1 |
NR_110615.1 |
Refseq |