Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01126089_m1
          See other NOL3 GE Assays ›
          Gene Symbol:
          NOL3
          Gene Name:
          nucleolar protein 3
          Gene Aliases:
          ARC, FCM, MYP, NOP, NOP30
          Chromosome Location:
          Chr.16: 67170497 - 67175737 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01126089_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NOL3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001276307.1 NP_001263236.1 4 - 5 521 114
          NM_001276309.1 NP_001263238.1 2 - 3 381 114
          NM_001276312.1 NP_001263241.1 4 - 5 872 114
          NM_001276319.1 NP_001263248.1 4 - 5 872 114
          NM_003946.6 NP_003937.1 2 - 3 497 114
          XM_017023843.1 XP_016879332.1 2 - 3 1134 114
          GenBank mRNA AF043244.1 - 2 - 3 414 114
          AF064600.1 - 2 - 3 372 114
          AK294145.1 - 4 - 5 837 114
          AK314899.1 - 2 - 3 402 114
          BC012798.2 - 2 - 3 383 3506011 114
          BP215200.1 - 2 - 3 338 114
          BQ685512.1 - 2 - 3 497 114
          BT020158.1 - 1 - 2 295 114
          CB959673.1 - 4 - 5 548 114

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NOL3

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          blood vessel remodeling
          response to ischemia
          mRNA splice site selection
          RNA splicing
          cardiac muscle cell apoptotic process
          negative regulation of cardiac muscle cell apoptotic process
          negative regulation of tumor necrosis factor-mediated signaling pathway
          negative regulation of muscle atrophy
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          response to injury involved in regulation of muscle adaptation
          negative regulation of apoptotic process
          protein complex oligomerization
          negative regulation of necrotic cell death
          negative regulation of release of cytochrome c from mitochondria
          intrinsic apoptotic signaling pathway
          regulation of NIK/NF-kappaB signaling
          negative regulation of mitochondrial membrane permeability involved in apoptotic process
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          inhibition of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of extrinsic apoptotic signaling pathway
          RNA binding
          death receptor binding
          calcium ion binding
          protein binding
          death effector domain binding
          identical protein binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          caspase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d58dc5f8-fsfvr:80/100.66.134.75:80.
          git-commit: 38b8a393e477ae4d9d89cc5f7a29948d21ae29e2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc