Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01127343_s1
          See other RASSF3 GE Assays ›
          Gene Symbol:
          RASSF3
          Gene Name:
          Ras association domain family member 3
          Gene Aliases:
          RASSF5
          Chromosome Location:
          Chr.12: 64610513 - 64697567 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01127343_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RASSF3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178169.3 NP_835463.1 5 2340 88
          NR_040718.1 4 2102 88
          XM_006719345.3 XP_006719408.1 5 2205 88
          XM_011538195.2 XP_011536497.1 5 2199 88
          XM_017019182.1 XP_016874671.1 5 2154 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF122970.1 - 1 179 Chr. 12 - 64696244 - 64696748 [+] on Build GRCh38 88
          GenBankmRNA CF122985.1 - 1 179 Chr. 12 - 64696244 - 64696953 [+] on Build GRCh38 88
          GenBankmRNA CF146884.1 - 1 229 Chr. 12 - 64696190 - 64696916 [+] on Build GRCh38 88
          GenBankmRNA CF145394.1 - 1 238 Chr. 12 - 64696184 - 64696869 [+] on Build GRCh38 88
          GenBankmRNA CF146865.1 - 1 229 Chr. 12 - 64696190 - 64696901 [+] on Build GRCh38 88
          GenBankmRNA CF135101.1 - 1 87 Chr. 12 - 64696336 - 64696897 [+] on Build GRCh38 88
          GenBankmRNA CF144851.1 - 1 214 Chr. 12 - 64696209 - 64696742 [+] on Build GRCh38 88
          GenBankmRNA BQ423046.1 - 1 69 Chr. 12 - 64696347 - 64697215 [+] on Build GRCh38 88
          GenBankmRNA BU177771.1 - 1 244 Chr. 12 - 64696172 - 64697009 [+] on Build GRCh38 88
          GenBankmRNA CF147156.1 - 1 308 Chr. 12 - 64696115 - 64696858 [+] on Build GRCh38 88
          GenBankmRNA AI989300.1 - 1 Chr. 12 - 64696347 - 64696816 [-] on Build GRCh38 88
          GenBankmRNA CF122199.1 - 1 213 Chr. 12 - 64696209 - 64696972 [+] on Build GRCh38 88
          GenBankmRNA CF142118.1 - 1 167 Chr. 12 - 64696252 - 64696739 [+] on Build GRCh38 88
          GenBankmRNA CF139886.1 - 1 214 Chr. 12 - 64696209 - 64696669 [+] on Build GRCh38 88
          GenBankmRNA AA303392.1 - 1 199 Chr. 12 - 64696217 - 64696500 [+] on Build GRCh38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 548z MIR548Z -72.2kb Anti-sense Chr. 12 - 64622509 - 64622605 on NCBI Build GRCh38
          Downstream glucosamine (N-acetyl)-6-sulfatase GNS G6S 15.9kb Anti-sense Chr. 12 - 64713442 - 64759446 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RASSF3

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          regulation of apoptotic process
          protein binding
          identical protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200011097.hg.1 ENST00000623666.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011097.hg.1 ENST00000623666.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 208499 - -
          ABI Human Genome Survey Microarray Version 2.0 234078 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline