Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01127792_m1
          See other MINPP1 GE Assays ›
          Gene Symbol:
          MINPP1
          Gene Name:
          multiple inositol-polyphosphate phosphatase 1
          Gene Aliases:
          HIPER1, MINPP2, MIPP
          Chromosome Location:
          Chr.10: 87502792 - 87553461 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01127792_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MINPP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001178118.1 NP_001171589.1 3 - 4 700 152
          NM_004897.4 NP_004888.2 3 - 4 1381 152
          XM_011540379.2 XP_011538681.1 4 - 5 1067 152
          XM_017016966.1 XP_016872455.1 4 - 5 1086 152
          GenBank mRNA AF046914.1 - 3 - 4 931 152
          AF084943.1 - 3 - 4 951 152
          AK304624.1 - 4 - 5 572 152
          AK309176.1 - 3 - 4 699 152
          AK312937.1 - 3 - 4 972 152
          AL050356.1 - 3 - 4 954 152
          AY358938.1 - 3 - 4 936 152
          BC032504.1 - 3 - 4 933 5243440 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008351.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000619.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MINPP1

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          polyphosphate metabolic process
          dephosphorylation
          bone mineralization
          inositol phosphate metabolic process
          acid phosphatase activity
          inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
          bisphosphoglycerate 3-phosphatase activity
          inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
          inositol hexakisphosphate 2-phosphatase activity
          protein histidine phosphatase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10006786.hg.1 ENST00000371996.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006786.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006786.hg.1 ENST00000546140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006786.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006786.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006786.hg.1 TC10000619.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 TC1000008351.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000156562.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371994.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000371996.8 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000472891.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 ENST00000536010.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178117.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_001178118.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq
          Affymetrix Human Clariom D Assay TC1000008351.hg.1 NM_004897.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0