Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01127825_g1
          See other HMGB2 GE Assays ›
          Gene Symbol:
          HMGB2
          Gene Name:
          high mobility group box 2
          Gene Aliases:
          HMG2
          Chromosome Location:
          Chr.4: 173331376 - 173334444 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01127825_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HMGB2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130688.1 NP_001124160.1 1 - 2 119 64
          GenBank mRNA AK311864.1 - 1 - 2 119 64
          BC100019.1 - 1 - 2 81 6163784 64

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK311864.1, BC100019.1, NM_001130688.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          inflammatory response to antigenic stimulus
          DNA topological change
          apoptotic DNA fragmentation
          chromatin organization
          nucleosome assembly
          transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          spermatid nucleus differentiation
          male gonad development
          response to xenobiotic stimulus
          positive regulation of nuclease activity
          DNA geometric change
          response to lipopolysaccharide
          positive regulation of interferon-beta production
          V(D)J recombination
          positive regulation of DNA binding
          innate immune response
          positive regulation of innate immune response
          positive regulation of erythrocyte differentiation
          positive regulation of megakaryocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          response to steroid hormone
          regulation of neurogenesis
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          positive chemotaxis
          DNA ligation involved in DNA repair
          cell chemotaxis
          cellular response to lipopolysaccharide
          regulation of stem cell proliferation
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          four-way junction DNA binding
          transcription cis-regulatory region binding
          DNA binding
          damaged DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          DNA-binding transcription factor activity
          RNA binding
          protein binding
          transcription factor binding
          DNA binding, bending
          protein domain specific binding
          chemoattractant activity
          non-sequence-specific DNA binding, bending
          RAGE receptor binding
          supercoiled DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900006517.hg.1 ENST00000591146.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 TC0400012432.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay PSR0400185202.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000296503.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000438704.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000446922.6 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000506267.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 ENST00000511316.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130688.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_001130689.1 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC0400012432.hg.1 NM_002129.3 Refseq
          Affymetrix Human Clariom D Assay TC1900006517.hg.1 ENST00000591146.2 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0