Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01240485_m1
          See other PPFIA2 GE Assays ›
          Gene Symbol:
          PPFIA2
          Gene Name:
          PTPRF interacting protein alpha 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 81257975 - 81759748 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01240485_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPFIA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001220473.2 NP_001207402.1 25 - 26 3320 65
          NM_001220474.2 NP_001207403.1 25 - 26 3275 65
          NM_001282536.1 NP_001269465.1 23 - 24 2827 65
          NM_003625.4 NP_003616.2 26 - 27 3183 65
          XM_006719670.3 XP_006719733.1 24 - 25 10384 65
          XM_017020079.1 XP_016875568.1 27 - 28 3176 65
          XM_017020080.1 XP_016875569.1 25 - 26 3314 65
          XM_017020084.1 XP_016875573.1 26 - 27 3323 65
          XM_017020086.1 XP_016875575.1 26 - 27 3167 65
          XM_017020088.1 XP_016875577.1 26 - 27 3556 65
          XM_017020089.1 XP_016875578.1 25 - 26 3269 65
          XM_017020094.1 XP_016875583.1 26 - 27 3101 65
          XM_017020096.1 XP_016875585.1 24 - 25 3239 65
          XM_017020099.1 XP_016875588.1 24 - 25 3260 65
          XM_017020105.1 XP_016875594.1 24 - 25 3239 65
          XM_017020110.1 XP_016875599.1 24 - 25 3472 65
          XM_017020112.1 XP_016875601.1 23 - 24 3139 65
          XM_017020114.1 XP_016875603.1 24 - 25 3148 65
          XM_017020115.1 XP_016875604.1 23 - 24 3139 65
          XM_017020117.1 XP_016875606.1 23 - 24 2883 65
          XM_017020118.1 XP_016875607.1 25 - 26 3123 65
          XM_017020120.1 XP_016875609.1 24 - 25 2836 65
          GenBank mRNA AB210009.1 - 25 - 26 3318 65
          AF034799.1 - 26 - 27 3191 65
          AK126971.1 - 25 - 26 3235 65
          AK302658.1 - 23 - 24 2798 65

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PPFIA2

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          neurotransmitter secretion
          glutamate secretion
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0