Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01369349_m1
          See other LOC647070 GE Assays ›
          Gene Symbol:
          LOC647070
          Gene Name:
          nascent polypeptide-associated complex subunit alpha, muscle-specific form-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 182833672 - 182839940 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01369349_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC647070
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK001442.1 - 1 - 2 280 79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AU128475.1 - 1 - 2 280 Chr. 1 - 182837404 - 182839591 [-] on Build GRCh38 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream N-acetylneuraminate pyruvate lyase NPL C112,
          C1orf13,
          NAL,
          NPL1
          -6.8kb Anti-sense Chr. 1 - 182789449 - 182830384 on NCBI Build GRCh38
          Downstream SHC binding and spindle associated 1 like SHCBP1L C1orf14,
          GE36
          60.3kb Sense Chr. 1 - 182899865 - 182953418 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3421219 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 51850 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3421219 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 215468_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 215467_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 215467_x_at - -
          Human Genome U133 Plus 2.0 Array 215467_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 215468_at - -
          Human Genome U133 Plus 2.0 Array 215468_PM_at - -
          Human Genome U133A 2.0 Array 215467_x_at - -
          Human Genome U133A 2.0 Array 215468_at - -
          Human Genome U133A 2.0 Array 215467_PM_x_at - -
          Human Genome U133A 2.0 Array 215468_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw528296 - -
          AffymetrixPathwork Diagnostics Pathchip pw571067 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215468_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215467_x_at8 - -
          Human Genome U95 Set 86748_r_at - -
          Human Genome U95 Set 86748_PM_r_at - -
          Human Genome U133 Set 215467_PM_x_at - -
          Human Genome U133 Set 215468_at - -
          Human Genome U133 Set 215468_PM_at - -
          Human Genome U133 Set 215467_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229967 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3286660 AK001442.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676285 AK001442.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676285 DB285127.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676285 DB223823.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676285 BC137136.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1676285 BC144494.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3286660 AK001442.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676285 AK001442.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676285 DB285127.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676285 DB223823.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676285 BC137136.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1676285 BC144494.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0