Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01369446_m1
          See other LOC100287877 GE Assays ›
          Gene Symbol:
          LOC100287877
          Gene Name:
          uncharacterized LOC100287877
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 5705568 - 5713772 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01369446_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100287877
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC039532.1 - 1 - 2 174 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM553100.1 - 194 Chr. - - [-] on Build 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 4417 MIR4417 mir-4417 -144.0kb Anti-sense Chr. 1 - 5564071 - 5564143 on NCBI Build GRCh38
          Downstream microRNA 4689 MIR4689 mir-4689 150.5kb Sense Chr. 1 - 5862672 - 5862741 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020465 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15017567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020449 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15017553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020479 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15017554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020485 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020499 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029467 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20029579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15017561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020488 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00020454 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 147442 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 858824 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 653446 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129835 AF501281.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129835 AK091270.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129835 AK128516.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129835 AK131360.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129835 AX746886.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129835 AY204751.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129835 NM_152701.2 refseq_mrna_human_NM
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00006486 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00006486 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00006486 - -
          Array Name Probe IDs Targets Target Type
          Human Genome U133 Plus 2.0 Array 1563008_PM_at - -
          Human Genome U133 Plus 2.0 Array 1563008_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563008_at4 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 695718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0