Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01370702_m1
          See other TDRD12 GE Assays ›
          Gene Symbol:
          TDRD12
          Gene Name:
          tudor domain containing 12
          Gene Aliases:
          ECAT8
          Chromosome Location:
          Chr.19: 32719773 - 32829580 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01370702_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TDRD12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110822.1 NP_001104292.1 7 - 8 1074 90
          XM_011527471.2 XP_011525773.1 7 - 8 834 90
          XM_011527472.2 XP_011525774.1 7 - 8 834 90
          XM_011527473.2 XP_011525775.1 7 - 8 834 90
          XM_011527474.2 XP_011525776.1 6 - 7 714 90
          XM_011527475.2 XP_011525777.1 7 - 8 834 90
          XM_011527476.2 XP_011525778.1 7 - 8 834 90
          XM_017027458.1 XP_016882947.1 7 - 8 834 90
          GenBank mRNA AB211063.1 - 6 - 7 756 90
          BC049000.1 - 7 - 8 1074 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900011705.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000430.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          male meiotic nuclear division
          multicellular organism development
          spermatogenesis
          biological_process
          fertilization
          RNA secondary structure unwinding
          cell differentiation
          gene silencing by RNA
          piRNA metabolic process
          DNA methylation involved in gamete generation
          positive regulation of protein targeting to mitochondrion
          molecular_function
          nucleic acid binding
          RNA helicase activity
          ATP binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19005583.hg.1 ENST00000421545.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005583.hg.1 ENST00000444215.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005583.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005583.hg.1 TC19000430.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011705.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900200247.hg.1 ENST00000421545.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900200247.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900200247.hg.1 TC1900011705.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900200247.hg.1 NM_001110822.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900200247.hg.1 ENST00000421545.2 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 126202 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0