Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
ENST00000457478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011537.hg.1 |
NR_110228.1 |
Refseq |