Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01379729_m1
          See other LOC158434 GE Assays ›
          Gene Symbol:
          LOC158434
          Gene Name:
          uncharacterized LOC158434
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 96106661 - 96116411 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01379729_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC158434
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_132344.1 1 - 2 163 67
          GenBank mRNA AK055173.1 - 1 - 2 164 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA434413.1 - 1 - 2 164 Chr. 9 - 96108804 - 96116411 [-] on Build GRCh38 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC158435 LOC158435 -12.7kb Anti-sense Chr. 9 - 96065839 - 96101912 on NCBI Build GRCh38
          Downstream hydroxysteroid 17-beta dehydrogenase 3 HSD17B3 EDH17B3,
          SDR12C2
          118.9kb Sense Chr. 9 - 96235306 - 96313987 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900010903.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175811 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833777 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833804 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833803 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833802 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833770 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27175808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16244247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06833764 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129845 AK127978.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129845 AK131483.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129845 AY162215.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129845 BC101092.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129845 BC101093.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129845 BC101094.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129845 BC101095.2 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129845 NM_181774.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 769425 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 918373 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 360791 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3776564 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43321 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P38802965 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3776564 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P38802965 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 126354 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16929738 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18702484 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16930173 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P16931752 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38802965 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P38802965 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18702484 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1563827_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174166.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174166.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174166.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174166.hg.1 TC0900010903.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174166.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174166.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174166.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174170.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174170.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174170.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174170.hg.1 TC0900010903.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174170.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174170.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174170.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay TC0900010903.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174167.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174167.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174167.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174167.hg.1 TC0900010903.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174167.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174167.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174167.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174169.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174169.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174169.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174169.hg.1 TC0900010903.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174169.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174169.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174169.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174168.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174168.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174168.hg.1 NR_132344.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900174168.hg.1 TC0900010903.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900174168.hg.1 ENST00000610562.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174168.hg.1 ENST00000624497.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900174168.hg.1 NR_132344.1 Refseq
          Human Genome U133 Plus 2.0 Array 1563827_PM_at - -
          Human Genome U133 Plus 2.0 Array 1563827_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563827_at2 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 698334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0