Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010340.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010340.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010340.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010340.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010340.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010340.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010340.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010340.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176442.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176442.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176442.hg.1 |
TC1400010340.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176442.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176442.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176446.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176446.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176446.hg.1 |
TC1400010340.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176446.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176446.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176444.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176444.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176444.hg.1 |
TC1400010340.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176444.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176444.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176443.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176443.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176443.hg.1 |
TC1400010340.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176443.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176443.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010340.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010340.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010340.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010340.hg.1 |
ENST00000557640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010340.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010340.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010340.hg.1 |
NM_001257268.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010340.hg.1 |
NM_001257268.1 |
Refseq |
Human Genome U95 Set |
69959_at |
- |
- |
Human Genome U95 Set |
69959_PM_at |
- |
- |