Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01383696_gH
          See other RFPL4A GE Assays ›
          Gene Symbol:
          RFPL4A RFPL4AL1
          Gene Name:
          ret finger protein like 4A
          ret finger protein like 4A like 1
          Gene Aliases:
          RFPL4, RNF210
          -
          Chromosome Location:
          Chr.19: 55757298 - 55763388 on Build GRCh38
          Chromosome Location:
          Chr.19: 55769118 - 55773423 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01383696_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RFPL4A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145014.1 NP_001138486.1 2 - 3 330 84
          XM_011526915.2 XP_011525217.1 3 - 4 1618 84
          Gene Symbol: RFPL4AL1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277397.1 NP_001264326.1 1 - 2 84
          XM_011527259.1 XP_011525561.1 2 - 3 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900008931.hg.1, TC1900008932.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000910.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 898734 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 132088 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 278168 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 57771 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 315568 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 267386 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 335975 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 272852 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 11123 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375790 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3366127 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1963 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1964 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43783 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 29644 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375790 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3366127 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3375790 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3366127 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647978 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647968 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003899 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647969 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003897 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647976 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647972 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003911 - -
          Agilent Whole Chr array_18 19 21 A_17_P17163530 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003903 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003907 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003902 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647974 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003914 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003906 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647973 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003896 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003908 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003900 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647971 - -
          Agilent Whole Chr array_18 19 21 A_17_P17163529 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647990 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003909 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647985 - -
          Agilent Whole Chr array_18 19 21 A_17_P31648012 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003894 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003910 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003898 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003905 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647979 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003895 - -
          Agilent Whole Chr array_18 19 21 A_17_P31648011 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003901 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003904 - -
          Agilent Whole Chr array_18 19 21 A_17_P17163505 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647970 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647973 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647985 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647990 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647971 - -
          Agilent Whole Chr array_18 19 21 A_17_P17163530 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003909 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647985 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647978 - -
          Agilent Whole Chr array_18 19 21 A_17_P31648011 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003905 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003911 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003901 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647971 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647976 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003914 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003904 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647973 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003897 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647968 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003908 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003902 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647979 - -
          Agilent Whole Chr array_18 19 21 A_17_P17163529 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003894 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647972 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647990 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003900 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003907 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647970 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003910 - -
          Agilent Whole Chr array_18 19 21 A_17_P17163505 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003899 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003898 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003903 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003895 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003906 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647974 - -
          Agilent Whole Chr array_18 19 21 A_17_P31648012 - -
          Agilent Whole Chr array_18 19 21 A_17_P31647969 - -
          Agilent Whole Chr array_18 19 21 A_17_P11003896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647971 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647990 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647973 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11003908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31647985 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11003903 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11003909 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11003894 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11003906 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11003896 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11003901 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11003897 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11003899 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8031574 - -
          Affymetrix Human Gene 1.0 ST Array 8031575 - -
          Affymetrix Human Gene 1.0 ST Array 8031571 - -
          Affymetrix Human Gene 1.0 ST Array 8031572 - -
          Affymetrix Human Gene 1.1 ST Array 8031570 - -
          Affymetrix Human Gene 1.1 ST Array 8031573 - -
          Affymetrix Human Genome U219 Array 11741830_PM_s_at - -
          Affymetrix Human Genome U219 Array 11741830_s_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1900008931.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008931.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008931.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008931.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008931.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008931.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008931.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19012577.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19012577.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19012577.hg.1 TC19000910.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900008932.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008932.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008932.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008932.hg.1 NM_001277397.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008932.hg.1 NM_001277397.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008933.hg.1 ENST00000530883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008933.hg.1 ENST00000530883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19012578.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19012578.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19012578.hg.1 TC19000910.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19012579.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19012579.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19012579.hg.1 TC19000910.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0400010239.hg.1 ENST00000515267.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010263.hg.1 ENST00000614131.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170117.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170117.hg.1 NM_001277397.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900170117.hg.1 TC1900008932.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900170117.hg.1 NM_001277397.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900170117.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170111.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170111.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900170111.hg.1 TC1900008931.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900170111.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170111.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008932.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900008932.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900008932.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900008932.hg.1 NM_001277397.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008932.hg.1 NM_001277397.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900170118.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170118.hg.1 NM_001277397.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900170118.hg.1 TC1900008932.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900170118.hg.1 ENST00000341750.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170118.hg.1 NM_001277397.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010263.hg.1 ENST00000614131.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008931.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008931.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008931.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008931.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008931.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008931.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Clariom D Assay TC1900008931.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900170113.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170113.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900170113.hg.1 TC1900008931.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900170113.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170113.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Clariom D Assay TC0400010239.hg.1 ENST00000515267.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008933.hg.1 ENST00000530883.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008933.hg.1 ENST00000530883.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170114.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170114.hg.1 NM_001145014.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900170114.hg.1 TC1900008931.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900170114.hg.1 ENST00000434937.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900170114.hg.1 NM_001145014.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013089 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013089 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 122462 - -
          ABI Human Genome Survey Microarray Version 2.0 492295 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1723341 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1723341 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1805439 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1805439 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0