Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01388458_m1
          See other LOC340107 GE Assays ›
          Gene Symbol:
          LOC340107
          Gene Name:
          uncharacterized LOC340107
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 24835280 - 24840583 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01388458_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC340107
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037896.1 2 - 3 420 93
          GenBank mRNA BC040315.1 - 2 - 3 420 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG773452.1 - 2 - 3 424 Chr. 5 - 24836865 - 24840585 [-] on Build GRCh38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cadherin 10 CDH10 -190.3kb Sense Chr. 5 - 24487100 - 24644978 on NCBI Build GRCh38
          Downstream uncharacterized LOC105374693 LOC105374693 351.7kb Anti-sense Chr. 5 - 25190953 - 25302280 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500010313.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001206.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15676239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824254 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824322 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994734 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994716 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994652 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824316 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994658 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824289 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824319 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994733 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994738 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23994648 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03824303 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 164992 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 190510 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 686700 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 503519 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3667419 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16262 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01198036 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01198036 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3667419 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3667419 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P01198036 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01198036 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 163368 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 47991 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17062762 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01198036 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01198036 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1557008_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1557009_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR05017123.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05017123.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05017123.hg.1 TC05001206.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05017125.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05017125.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05017125.hg.1 TC05001206.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05017124.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05017124.hg.1 NR_037896.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05017124.hg.1 TC05001206.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500171906.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171906.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171906.hg.1 TC0500010313.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171906.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171906.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171911.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171911.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171911.hg.1 TC0500010313.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171911.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171911.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay TC0500010313.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171905.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171905.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171905.hg.1 TC0500010313.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171905.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171905.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171907.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171907.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171907.hg.1 TC0500010313.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171907.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171907.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171910.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171910.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171910.hg.1 TC0500010313.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171910.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171910.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171909.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171909.hg.1 NR_037896.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500171909.hg.1 TC0500010313.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500171909.hg.1 ENST00000504339.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500171909.hg.1 NR_037896.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557009_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1557008_PM_at - -
          Human Genome U133 Plus 2.0 Array 1557009_a_at - -
          Human Genome U133 Plus 2.0 Array 1557008_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557009_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557008_at8 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A026614 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A026614 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 245704 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1840923 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 6900241 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_78165 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1840923 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1840923 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1840923 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0