Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022064.4 | NP_071347.2 | 25 - 26 | 2535 | 73 | |
NR_135218.1 | 25 - 26 | 2539 | 73 | |||
XM_006713285.1 | XP_006713348.1 | 25 - 26 | 2693 | 73 | ||
XM_011533995.1 | XP_011532297.1 | 25 - 26 | 2470 | 73 | ||
XM_017007018.1 | XP_016862507.1 | 25 - 26 | 2497 | 73 | ||
GenBank mRNA | AF370367.1 | - | 16 - 17 | 1689 | 73 | |
AK022627.1 | - | 3 - 4 | 361 | 73 | ||
AK026968.1 | - | 7 - 8 | 789 | 73 | ||
AK090632.1 | - | 15 - 16 | 1577 | 73 | ||
AK096353.1 | - | 25 - 26 | 2449 | 73 | ||
AK126883.1 | - | 10 - 11 | 1386 | 73 | ||
AK308607.1 | - | 22 - 23 | 2213 | 73 | ||
AL832138.1 | - | 24 - 25 | 2703 | 73 | ||
AL832344.1 | - | 25 - 26 | 2474 | 73 | ||
AY744152.1 | - | 24 - 25 | 2418 | 73 | ||
BC041145.1 | - | 10 - 11 | 1068 | 73 | ||
BC057392.1 | - | 12 - 13 | 1225 | 73 | ||
BC088801.1 | - | 7 - 8 | 796 | 73 | ||
BC130632.1 | - | 25 - 26 | 2476 | 40146663 | 73 | |
CR627051.1 | - | 22 - 23 | 3251 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |