Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01547991_m1
See other ANK2 GE Assays ›
Gene Symbol:
ANK2
Gene Name:
ankyrin 2, neuronal
Gene Aliases:
ANK-2, LQT4, brank-2
Chromosome Location:
Chr.4: 112706083 - 113383740 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01547991_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ANK2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001127493.1
NP_001120965.1
38 - 39
4591
136
NM_020977.3
NP_066187.2
36 - 37
4497
136
XM_017008078.1
XP_016863567.1
36 - 37
4595
136
XM_017008079.1
XP_016863568.1
34 - 35
4325
136
XM_017008080.1
XP_016863569.1
36 - 37
4446
136
XM_017008081.1
XP_016863570.1
35 - 36
4496
136
XM_017008082.1
XP_016863571.1
37 - 38
4673
136
XM_017008083.1
XP_016863572.1
37 - 38
4736
136
XM_017008084.1
XP_016863573.1
37 - 38
4631
136
XM_017008085.1
XP_016863574.1
37 - 38
4592
136
XM_017008086.1
XP_016863575.1
36 - 37
4493
136
XM_017008087.1
XP_016863576.1
38 - 39
4772
136
XM_017008088.1
XP_016863577.1
39 - 40
4566
136
XM_017008089.1
XP_016863578.1
36 - 37
4739
136
XM_017008090.1
XP_016863579.1
37 - 38
4736
136
XM_017008091.1
XP_016863580.1
35 - 36
4433
136
XM_017008092.1
XP_016863581.1
37 - 38
4533
136
XM_017008093.1
XP_016863582.1
38 - 39
4530
136
XM_017008094.1
XP_016863583.1
37 - 38
4592
136
XM_017008095.1
XP_016863584.1
35 - 36
4410
136
XM_017008096.1
XP_016863585.1
39 - 40
4566
136
XM_017008097.1
XP_016863586.1
36 - 37
4371
136
XM_017008098.1
XP_016863587.1
37 - 38
4533
136
XM_017008099.1
XP_016863588.1
38 - 39
4530
136
XM_017008100.1
XP_016863589.1
35 - 36
4335
136
XM_017008101.1
XP_016863590.1
35 - 36
4496
136
XM_017008102.1
XP_016863591.1
34 - 35
4311
136
XM_017008103.1
XP_016863592.1
37 - 38
4592
136
XM_017008104.1
XP_016863593.1
36 - 37
4434
136
XM_017008105.1
XP_016863594.1
35 - 36
4410
136
XM_017008106.1
XP_016863595.1
34 - 35
4397
136
XM_017008107.1
XP_016863596.1
36 - 37
4371
136
XM_017008108.1
XP_016863597.1
34 - 35
4295
136
XM_017008110.1
XP_016863599.1
36 - 37
4434
136
XM_017008111.1
XP_016863600.1
35 - 36
4410
136
XM_017008112.1
XP_016863601.1
33 - 34
4212
136
GenBank mRNA
AK294720.1
-
15 - 16
2242
136
AK299893.1
-
9 - 10
1339
136
BC125235.1
-
4 - 5
382
136
BC125236.1
-
4 - 5
382
136
BX537758.1
-
38 - 39
4565
136
X56958.2
-
36 - 37
4412
136
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact
Tech Support
if you need more information.
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ANK2
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
atrial septum development
cellular calcium ion homeostasis
endoplasmic reticulum to Golgi vesicle-mediated transport
cytoskeleton organization
positive regulation of gene expression
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of cardiac muscle contraction by calcium ion signaling
paranodal junction assembly
regulation of protein stability
T-tubule organization
protein localization to organelle
protein localization to cell surface
cellular protein localization
protein localization to M-band
protein localization to T-tubule
positive regulation of potassium ion transport
protein stabilization
regulation of release of sequestered calcium ion into cytosol
response to methylmercury
regulation of calcium ion transport
positive regulation of calcium ion transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
sarcoplasmic reticulum calcium ion transport
protein localization to endoplasmic reticulum
protein localization to plasma membrane
regulation of cardiac muscle cell contraction
ventricular cardiac muscle cell action potential
atrial cardiac muscle cell action potential
SA node cell action potential
membrane depolarization during SA node cell action potential
atrial cardiac muscle cell to AV node cell communication
SA node cell to atrial cardiac muscle cell communication
regulation of heart rate by cardiac conduction
regulation of SA node cell action potential
regulation of atrial cardiac muscle cell action potential
positive regulation of potassium ion transmembrane transporter activity
regulation of calcium ion transmembrane transporter activity
positive regulation of calcium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal anchor activity
enzyme binding
protein kinase binding
spectrin binding
protein-macromolecule adaptor activity
transmembrane transporter binding
ATPase binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0