Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01548406_m1
          See other FAR1 GE Assays ›
          Gene Symbol:
          FAR1
          Gene Name:
          fatty acyl-CoA reductase 1
          Gene Aliases:
          MLSTD2, PFCRD, SDR10E1
          Chromosome Location:
          Chr.11: 13668659 - 13732346 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01548406_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FAR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032228.5 NP_115604.1 2 - 3 341 106
          XM_011520400.1 XP_011518702.1 2 - 3 341 106
          GenBank mRNA AK026381.1 - 2 - 3 289 106
          AK315527.1 - 2 - 3 290 106
          AY358784.1 - 2 - 3 330 106
          AY423606.1 - 2 - 3 284 106
          AY600449.1 - 2 - 3 276 106
          BC017377.2 - 2 - 3 283 3835094 106
          CR936619.1 - 2 - 3 316 106
          DA515509.1 - 2 - 3 341 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100006906.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000208.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FAR1

          Gene Ontology Categories:

          Function(s) Process(es)

          ether lipid biosynthetic process
          wax biosynthetic process
          long-chain fatty-acyl-CoA metabolic process
          glycerophospholipid biosynthetic process
          oxidoreductase activity
          long-chain-fatty-acyl-CoA reductase activity
          fatty-acyl-CoA reductase (alcohol-forming) activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11002807.hg.1 ENST00000354817.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11002807.hg.1 ENST00000355107.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11002807.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11002807.hg.1 TC11000208.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 TC1100006906.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148620.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 TC1100006906.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100148621.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000354817.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000524933.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000527202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532502.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532701.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 ENST00000532769.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq
          Affymetrix Human Clariom D Assay TC1100006906.hg.1 NM_032228.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0