Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030435.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030435.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030435.hg.1 |
TC01001960.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100189337.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189337.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189337.hg.1 |
TC0100012123.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100189337.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189337.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
ENST00000450451.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012123.hg.1 |
NR_027247.2 |
Refseq |