Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01550837_m1
          See other STK26 GE Assays ›
          Gene Symbol:
          STK26
          Gene Name:
          serine/threonine protein kinase 26
          Gene Aliases:
          MASK, MST4
          Chromosome Location:
          Chr.X: 132023217 - 132075943 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01550837_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: STK26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042453.1 NP_001035918.1 6 - 7 854 137
          NM_016542.3 NP_057626.2 7 - 8 1085 137
          XM_011531349.1 XP_011529651.1 8 - 9 1058 137
          XM_011531350.1 XP_011529652.1 7 - 8 1131 137
          GenBank mRNA AB040057.2 - 7 - 8 1004 137
          AF231012.1 - 7 - 8 973 137
          AF344882.1 - 6 - 7 794 137
          AK000556.1 - 3 - 4 417 137
          AK027589.1 - 2 - 3 302 137
          AK074837.1 - 2 - 3 338 137
          AK075107.1 - 6 - 7 766 137
          AK075150.1 - 7 - 8 1000 137
          AK303583.1 - 8 - 9 1066 137
          AK314356.1 - 7 - 8 1001 137
          AK316137.1 - 8 - 9 1066 137
          BC098315.1 - 7 - 8 990 40026349 137
          BC099843.1 - 7 - 8 990 40026348 137
          BC103503.1 - 7 - 8 990 40026350 137
          BT020099.1 - 6 - 7 784 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00008437.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000628.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene STK26

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          protein phosphorylation
          cellular component disassembly involved in execution phase of apoptosis
          microvillus assembly
          response to hydrogen peroxide
          regulation of apoptotic process
          regulation of cell differentiation
          protein autophosphorylation
          regulation of hydrogen peroxide-induced cell death
          magnesium ion binding
          protein kinase activity
          protein binding
          ATP binding
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 ENST00000354719.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 ENST00000394334.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 ENST00000394335.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 ENST00000481105.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 ENST00000496850.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 NM_001042452.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 NM_001042453.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 NM_016542.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X008304.hg.1 TC0X000628.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000354719.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000394334.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000394335.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000481105.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000496850.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 NM_001042452.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 NM_001042453.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 NM_016542.3 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 TC0X00008437.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000354719.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000394334.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000394335.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000481105.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 ENST00000496850.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 NM_001042452.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 NM_001042453.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00157430.hg.1 NM_016542.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0