Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01551992_m1
          See other F7 GE Assays ›
          Gene Symbol:
          F7
          Gene Name:
          coagulation factor VII
          Gene Aliases:
          SPCA
          Chromosome Location:
          Chr.13: 113105773 - 113120681 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01551992_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000131.4 NP_000122.1 6 - 7 623 137
          NM_001267554.1 NP_001254483.1 3 - 4 371 137
          NM_019616.3 NP_062562.1 5 - 6 557 137
          NR_051961.1 4 - 5 590 137
          XM_011537474.2 XP_011535776.2 6 - 7 680 137
          XM_011537475.2 XP_011535777.2 4 - 5 494 137
          XM_011537476.2 XP_011535778.1 3 - 4 1038 137
          GenBank mRNA AK298404.1 - 3 - 4 368 137
          AK310779.1 - 5 - 6 557 137
          AK311565.1 - 5 - 6 472 137
          AK311682.1 - 4 - 5 539 137
          BC130468.1 - 5 - 6 547 40146499 137
          EU557239.1 - 6 - 7 573 137
          M13232.1 - 6 - 7 604 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300008108.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000416.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene F7

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of leukocyte chemotaxis
          signal peptide processing
          ER to Golgi vesicle-mediated transport
          blood coagulation
          blood coagulation, extrinsic pathway
          circadian rhythm
          positive regulation of platelet-derived growth factor receptor signaling pathway
          response to organic cyclic compound
          protein processing
          peptidyl-glutamic acid carboxylation
          positive regulation of blood coagulation
          positive regulation of cell migration
          organ regeneration
          response to vitamin K
          response to estrogen
          positive regulation of positive chemotaxis
          positive regulation of protein kinase B signaling
          response to growth hormone
          glycoprotein binding
          serine-type endopeptidase activity
          receptor binding
          calcium ion binding
          protein binding
          serine-type peptidase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000346342.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000375581.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000444337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000444337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000444337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000444337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000444337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000473085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000473085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000473085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000473085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000479674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000479674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000479674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000479674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000479674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 ENST00000541084.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_000131.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_001267554.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NM_019616.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008108.hg.1 NR_051961.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0