Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01552601_m1
          See other DNM1L GE Assays ›
          Gene Symbol:
          DNM1L
          Gene Name:
          dynamin 1 like
          Gene Aliases:
          DLP1, DRP1, DVLP, DYMPLE, EMPF, HDYNIV
          Chromosome Location:
          Chr.12: 32679200 - 32745650 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01552601_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DNM1L
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005690.4 NP_005681.2 14 - 15 1762 89
          XM_005253282.4 XP_005253339.1 15 - 16 1773 89
          XM_017018665.1 XP_016874154.1 13 - 14 1479 89
          GenBank mRNA AF061795.1 - 14 - 15 1619 89
          AK291094.1 - 14 - 15 1642 89
          BC107763.1 - 14 - 15 1646 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200007277.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DNM1L

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial fission
          release of cytochrome c from mitochondria
          dynamin family protein polymerization involved in mitochondrial fission
          endocytosis
          cellular component disassembly involved in execution phase of apoptosis
          regulation of mitochondrion organization
          peroxisome fission
          regulation of protein oligomerization
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          mitochondrial fragmentation involved in apoptotic process
          positive regulation of protein secretion
          protein complex oligomerization
          protein homotetramerization
          heart contraction
          membrane fusion
          necroptotic process
          mitochondrion morphogenesis
          protein localization to mitochondrion
          positive regulation of mitochondrial fission
          mitochondrial membrane fission
          positive regulation of release of cytochrome c from mitochondria
          regulation of peroxisome organization
          regulation of autophagy of mitochondrion
          regulation of ATP metabolic process
          positive regulation of intrinsic apoptotic signaling pathway
          GTPase activity
          GTPase activator activity
          protein binding
          GTP binding
          lipid binding
          GTP-dependent protein binding
          ubiquitin protein ligase binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000266481.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000358214.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000381000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000414834.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000452533.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000546649.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000546757.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000547312.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000547640.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000549157.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000549701.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000553031.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000553257.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_001278463.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_001278464.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_001278465.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_001278466.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_005690.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_012062.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_012063.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 TC1200007277.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000266481.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000358214.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000381000.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000414834.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000452533.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000546649.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000546757.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000547312.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000547640.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000549157.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000549701.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000553031.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 ENST00000553257.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_001278463.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_001278464.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_001278465.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_001278466.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_005690.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_012062.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200151099.hg.1 NM_012063.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0