Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000264052.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000340126.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000341950.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000409112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000409341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000409824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000409897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000414648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
ENST00000427101.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
NM_001080391.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
NM_001206701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
NM_001206702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
NM_001206703.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
NM_001206704.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
NM_003113.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02021556.hg.1 |
TC02001377.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000264052.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000340126.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000409112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000409341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000409824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000409897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000427101.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000432979.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001080391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001206701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001206702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001206703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001206704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_003113.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
TC0200011023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000409341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000264052.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000340126.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000409112.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000409824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000409897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000427101.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
ENST00000432979.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001080391.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001206701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001206702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001206703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_001206704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200179046.hg.1 |
NM_003113.3 |
Refseq |