Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080398.1 | NP_001073867.1 | 42 - 43 | 4869 | 92 | |
XM_005251850.3 | XP_005251907.1 | 40 - 41 | 4372 | 92 | ||
XM_005251851.4 | XP_005251908.1 | 41 - 42 | 4751 | 92 | ||
XM_005251852.4 | XP_005251909.1 | 41 - 42 | 4703 | 92 | ||
XM_005251853.3 | XP_005251910.1 | 41 - 42 | 4485 | 92 | ||
XM_005251854.3 | XP_005251911.1 | 40 - 41 | 4654 | 92 | ||
XM_011518422.2 | XP_011516724.1 | 40 - 41 | 4372 | 92 | ||
XM_017014557.1 | XP_016870046.1 | 34 - 35 | 3691 | 92 | ||
GenBank mRNA | AB002366.1 | - | 30 - 31 | 3143 | 92 | |
AK024705.1 | - | 17 - 18 | 1698 | 92 | ||
AK025689.1 | - | 25 - 26 | 2671 | 92 | ||
AK027063.1 | - | 10 - 11 | 839 | 92 | ||
AK127247.1 | - | 36 - 37 | 3810 | 92 | ||
BC021127.1 | - | 6 - 7 | 453 | 92 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |