Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01562006_g1
          See other TTN GE Assays ›
          Gene Symbol:
          TTN
          Gene Name:
          titin
          Gene Aliases:
          CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD
          Chromosome Location:
          Chr.2: 178525989 - 178807423 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01562006_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TTN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256850.1 NP_001243779.1 19 - 20 3390 99
          NM_001267550.2 NP_001254479.2 19 - 20 3390 99
          NM_003319.4 NP_003310.4 18 - 19 3252 99
          NM_133378.4 NP_596869.4 19 - 20 3390 99
          NM_133379.4 NP_596870.2 19 - 20 3390 99
          NM_133432.3 NP_597676.3 18 - 19 3252 99
          NM_133437.4 NP_597681.4 18 - 19 3252 99
          XM_017004819.1 XP_016860308.1 19 - 20 3393 99
          XM_017004820.1 XP_016860309.1 19 - 20 3393 99
          XM_017004821.1 XP_016860310.1 19 - 20 3390 99
          XM_017004822.1 XP_016860311.1 19 - 20 3393 99
          XM_017004823.1 XP_016860312.1 19 - 20 3393 99
          GenBank mRNA X90568.1 - 18 - 19 3159 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200016752.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02005047.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TTN

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          cardiac muscle hypertrophy
          muscle contraction
          striated muscle contraction
          mitotic chromosome condensation
          peptidyl-tyrosine phosphorylation
          muscle filament sliding
          skeletal muscle thin filament assembly
          skeletal muscle myosin thick filament assembly
          detection of muscle stretch
          sarcomere organization
          regulation of protein kinase activity
          sarcomerogenesis
          regulation of catalytic activity
          response to calcium ion
          cardiac myofibril assembly
          cardiac muscle tissue morphogenesis
          cardiac muscle cell development
          cardiac muscle contraction
          protease binding
          protein serine/threonine kinase activity
          protein tyrosine kinase activity
          calcium ion binding
          protein binding
          calmodulin binding
          ATP binding
          structural constituent of muscle
          enzyme binding
          protein kinase binding
          telethonin binding
          identical protein binding
          actinin binding
          protein self-association
          actin filament binding
          muscle alpha-actinin binding
          structural molecule activity conferring elasticity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 ENST00000342175.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 ENST00000342992.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 ENST00000356127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 ENST00000359218.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 ENST00000360870.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 ENST00000460472.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 NM_003319.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 NM_133378.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 NM_133379.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 NM_133432.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 NM_133437.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02041253.hg.1 TC02005047.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000342175.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000342992.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000359218.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000360870.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000460472.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000589042.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000591111.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000615779.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_001256850.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_001267550.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_003319.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_133378.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_133379.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_133432.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_133437.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000360870.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 TC0200016752.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000342175.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000342992.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000359218.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000460472.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000589042.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000591111.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 ENST00000615779.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_001256850.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_001267550.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_003319.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_133378.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_133379.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_133432.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200228498.hg.1 NM_133437.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0