Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01564979_g1
          See other SFI1 GE Assays ›
          Gene Symbol:
          SFI1
          Gene Name:
          SFI1 centrin binding protein
          Gene Aliases:
          PISD, PPP1R139, hSfi1p
          Chromosome Location:
          Chr.22: 31496139 - 31618554 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01564979_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SFI1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007467.2 NP_001007468.1 28 - 29 3598 120
          NM_001258325.1 NP_001245254.1 26 - 27 3433 120
          NM_001258326.1 NP_001245255.1 26 - 27 3348 120
          NM_001258327.1 NP_001245256.1 26 - 27 3316 120
          NM_014775.3 NP_055590.2 27 - 28 3505 120
          XM_005261868.2 XP_005261925.1 28 - 29 3596 120
          XM_006724390.2 XP_006724453.1 26 - 27 3350 120
          XM_011530574.2 XP_011528876.1 29 - 30 3324 120
          XM_011530581.2 XP_011528883.1 29 - 30 3324 120
          XM_017029141.1 XP_016884630.1 28 - 29 3231 120
          XM_017029144.1 XP_016884633.1 27 - 28 3503 120
          XM_017029145.1 XP_016884634.1 26 - 27 3431 120
          XM_017029149.1 XP_016884638.1 27 - 28 3524 120
          XM_017029150.1 XP_016884639.1 27 - 28 3520 120
          XM_017029151.1 XP_016884640.1 27 - 28 3499 120
          XM_017029154.1 XP_016884643.1 26 - 27 3427 120
          XM_017029155.1 XP_016884644.1 25 - 26 3257 120
          XM_017029156.1 XP_016884645.1 25 - 26 3257 120
          XM_017029162.1 XP_016884651.1 24 - 25 2858 120
          GenBank mRNA AB011114.2 - 27 - 28 3369 120
          AK055147.1 - 16 - 17 1837 120
          AK292408.1 - 28 - 29 3174 120
          AK293987.1 - 26 - 27 3046 120
          AK303362.1 - 26 - 27 2947 120
          BC021576.1 - 10 - 11 1165 120
          BC110814.1 - 26 - 27 2949 6500257 120
          BC129926.1 - 24 - 25 2891 40124134 120
          BC129945.1 - 22 - 23 2597 40124130 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200009261.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22001423.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SFI1

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          negative regulation of phosphatase activity
          protein binding
          phosphatase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 TC22001423.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000400288.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000400289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000414585.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000421060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000432498.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000443011.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000443326.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000474741.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 ENST00000540643.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 NM_001007467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22004404.hg.1 NM_014775.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000357852.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000382162.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000400288.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000400289.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000432498.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000443011.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000452250.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000464333.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000474741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000476577.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000540643.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_001007467.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_001258325.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_001258326.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_001258327.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_014775.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 TC2200009261.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000357852.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000382162.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000400288.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000400289.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000432498.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000443011.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000452250.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000464333.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000474741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000476577.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 ENST00000540643.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_001007467.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_001258325.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_001258326.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_001258327.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200168728.hg.1 NM_014775.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0