Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003847.hg.1 |
ENST00000005257.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003847.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003847.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003847.hg.1 |
TC07000250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000436179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000436179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000436179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000436179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003850.hg.1 |
ENST00000005257.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003850.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003850.hg.1 |
TC07000250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003849.hg.1 |
ENST00000005257.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003849.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003849.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003849.hg.1 |
TC07000250.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000436179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000436179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000436179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000436179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007271.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
TC0700007271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150452.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
TC0700007271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
ENST00000434466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
ENST00000466491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150453.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150455.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150455.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150455.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150455.hg.1 |
TC0700007271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700150455.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150455.hg.1 |
ENST00000468201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150455.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150456.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150456.hg.1 |
NM_005402.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150456.hg.1 |
TC0700007271.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700150456.hg.1 |
ENST00000005257.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150456.hg.1 |
NM_005402.3 |
Refseq |