Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01566267_m1
          See other MEF2C GE Assays ›
          Gene Symbol:
          MEF2C
          Gene Name:
          myocyte enhancer factor 2C
          Gene Aliases:
          C5DELq14.3, DEL5q14.3
          Chromosome Location:
          Chr.5: 88718241 - 88904105 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01566267_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MEF2C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001193349.1 NP_001180278.1 6 - 7 822 76
          NM_001193350.1 NP_001180279.1 8 - 9 1478 76
          NM_002397.4 NP_002388.2 8 - 9 1481 76
          XM_005248511.2 XP_005248568.1 8 - 9 1015 76
          XM_006714619.2 XP_006714682.1 9 - 10 1337 76
          XM_011543396.2 XP_011541698.1 8 - 9 1066 76
          XM_011543397.2 XP_011541699.1 8 - 9 1247 76
          XM_011543400.1 XP_011541702.1 8 - 9 1253 76
          XM_017009475.1 XP_016864964.1 8 - 9 1308 76
          XM_017009479.1 XP_016864968.1 8 - 9 1314 76
          XM_017009480.1 XP_016864969.1 9 - 10 1337 76
          XM_017009481.1 XP_016864970.1 8 - 9 1015 76
          GenBank mRNA AK307883.1 - 7 - 8 823 76
          AK312472.1 - 8 - 9 1186 76
          BC026341.1 - 8 - 9 1253 76
          FM180475.1 - 6 - 7 683 76
          L08895.1 - 8 - 9 1228 76

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MEF2C

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          MAPK cascade
          blood vessel development
          osteoblast differentiation
          neuron migration
          B cell homeostasis
          heart looping
          endochondral ossification
          blood vessel remodeling
          chondrocyte differentiation
          germinal center formation
          regulation of germinal center formation
          primary heart field specification
          secondary heart field specification
          outflow tract morphogenesis
          sinoatrial valve morphogenesis
          cardiac ventricle formation
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          apoptotic process
          humoral immune response
          nervous system development
          heart development
          muscle organ development
          skeletal muscle tissue development
          muscle cell fate determination
          learning or memory
          response to virus
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of alkaline phosphatase activity
          neural crest cell differentiation
          cardiac muscle hypertrophy in response to stress
          myotube differentiation
          neuron differentiation
          platelet formation
          monocyte differentiation
          negative regulation of ossification
          melanocyte differentiation
          positive regulation of bone mineralization
          positive regulation of B cell proliferation
          skeletal muscle cell differentiation
          cellular response to trichostatin A
          B cell proliferation
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          regulation of megakaryocyte differentiation
          positive regulation of myoblast differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of neurotransmitter secretion
          regulation of synaptic plasticity
          positive regulation of skeletal muscle tissue development
          neuron development
          cell morphogenesis involved in neuron differentiation
          embryonic viscerocranium morphogenesis
          negative regulation of epithelial cell proliferation
          B cell receptor signaling pathway
          smooth muscle cell differentiation
          positive regulation of muscle cell differentiation
          regulation of synapse assembly
          regulation of synaptic transmission, glutamatergic
          ventricular cardiac muscle cell differentiation
          roof of mouth development
          regulation of synaptic activity
          positive regulation of cardiac muscle cell proliferation
          excitatory postsynaptic potential
          regulation of sarcomere organization
          cartilage morphogenesis
          regulation of dendritic spine development
          renal tubule morphogenesis
          cellular response to lipopolysaccharide
          cellular response to calcium ion
          cellular response to parathyroid hormone stimulus
          cellular response to xenobiotic stimulus
          cellular response to fluid shear stress
          cellular response to transforming growth factor beta stimulus
          positive regulation of cell proliferation in bone marrow
          glomerulus morphogenesis
          nephron tubule epithelial cell differentiation
          positive regulation of protein homodimerization activity
          positive regulation of macrophage apoptotic process
          regulation of NMDA receptor activity
          regulation of AMPA receptor activity
          positive regulation of cardiac muscle cell differentiation
          positive regulation of behavioral fear response
          epithelial cell proliferation involved in renal tubule morphogenesis
          positive regulation of skeletal muscle cell differentiation
          transcription cis-regulatory region binding
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          core promoter sequence-specific DNA binding
          minor groove of adenine-thymine-rich DNA binding
          chromatin binding
          DNA-binding transcription factor activity
          protein binding
          miRNA binding
          histone deacetylase binding
          protein heterodimerization activity
          HMG box domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0