Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261463.1 | NP_001248392.1 | 12 - 13 | 1395 | 79 | |
NM_001322945.1 | NP_001309874.1 | 12 - 13 | 1395 | 79 | ||
NM_198060.3 | NP_932326.2 | 12 - 13 | 1395 | 79 | ||
XM_005269865.2 | XP_005269922.1 | 12 - 13 | 1371 | 79 | ||
XM_005269867.2 | XP_005269924.1 | 12 - 13 | 1371 | 79 | ||
XM_006717870.2 | XP_006717933.1 | 12 - 13 | 1371 | 79 | ||
XM_011539832.2 | XP_011538134.1 | 12 - 13 | 1371 | 79 | ||
GenBank mRNA | AK056605.1 | - | 4 - 5 | 405 | 79 | |
AK056969.1 | - | 12 - 13 | 1372 | 79 | ||
AL831980.2 | - | 12 - 13 | 1322 | 79 | ||
AL832025.2 | - | 12 - 13 | 1322 | 79 | ||
AY177620.1 | - | 12 - 13 | 1348 | 79 | ||
BC126407.1 | - | 12 - 13 | 1231 | 8992123 | 79 | |
BC143751.1 | - | 12 - 13 | 1231 | 9052265 | 79 | |
BX640682.2 | - | 12 - 13 | 1322 | 79 | ||
BX640730.1 | - | 12 - 13 | 1479 | 79 | ||
BX641052.1 | - | 12 - 13 | 1359 | 79 | ||
BX647658.1 | - | 12 - 13 | 1396 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |