Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01568043_m1
          See other ZFP36L1 GE Assays ›
          Gene Symbol:
          ZFP36L1
          Gene Name:
          ZFP36 ring finger protein like 1
          Gene Aliases:
          BRF1, Berg36, ERF-1, ERF1, RNF162B, TIS11B, cMG1
          Chromosome Location:
          Chr.14: 68787655 - 68796243 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01568043_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZFP36L1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001244698.1 NP_001231627.1 2 - 3 2002 136
          GenBank mRNA AK024202.1 - 2 - 3 2826 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400009524.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14001253.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          vasculogenesis
          proepicardium development
          regulation of transcription, DNA-templated
          apoptotic process
          heart development
          cell population proliferation
          response to wounding
          regulation of gene expression
          regulation of keratinocyte proliferation
          phosphatidylinositol 3-kinase signaling
          neural tube development
          nuclear-transcribed mRNA catabolic process, deadenylation-independent decay
          regulation of mRNA 3'-end processing
          cellular response to insulin stimulus
          T cell differentiation in thymus
          multicellular organism growth
          p38MAPK cascade
          regulation of mRNA stability
          protein kinase B signaling
          cellular response to fibroblast growth factor stimulus
          positive regulation of fat cell differentiation
          regulation of keratinocyte differentiation
          negative regulation of erythrocyte differentiation
          positive regulation of monocyte differentiation
          regulation of myoblast differentiation
          mesendoderm development
          mRNA transport
          chorio-allantoic fusion
          spongiotrophoblast layer development
          3'-UTR-mediated mRNA destabilization
          ERK1 and ERK2 cascade
          cellular response to cAMP
          cellular response to tumor necrosis factor
          cellular response to epidermal growth factor stimulus
          cellular response to peptide hormone stimulus
          cellular response to glucocorticoid stimulus
          cellular response to hypoxia
          cellular response to salt stress
          cellular response to transforming growth factor beta stimulus
          regulation of stem cell proliferation
          cellular response to raffinose
          positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          regulation of keratinocyte apoptotic process
          positive regulation of intracellular mRNA localization
          cellular response to phorbol 13-acetate 12-myristate
          DNA binding
          DNA-binding transcription factor activity
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          protein binding
          mRNA 3'-UTR AU-rich region binding
          metal ion binding
          14-3-3 protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14014219.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14014219.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14014219.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14014219.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14014219.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14014219.hg.1 TC14001253.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000553375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000553375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000553375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000553375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000557022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000557022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000557022.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000557086.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000557086.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000557086.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 ENST00000557086.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 TC1400009524.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170214.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 TC1400009524.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400170215.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000336440.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000439696.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000553375.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000553375.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000553375.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000553375.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000555997.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000557022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000557022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000557022.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000557086.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000557086.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000557086.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 ENST00000557086.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244698.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_001244701.1 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Affymetrix Human Clariom D Assay TC1400009524.hg.1 NM_004926.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 160743 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0