Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018355.hg.1 |
ENST00000296803.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018355.hg.1 |
ENST00000399564.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018355.hg.1 |
ENST00000506092.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018355.hg.1 |
NM_173489.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018355.hg.1 |
TC05001303.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
ENST00000399564.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
ENST00000503890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
ENST00000506092.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
ENST00000515297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
NM_173489.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
TC0500010591.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
ENST00000399564.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
ENST00000503890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
ENST00000506092.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
ENST00000515297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173907.hg.1 |
NM_173489.4 |
Refseq |