Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000426753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000426753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000428772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000428772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000439507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000439507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000457508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
NR_109916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
NR_109917.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
NR_109918.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
TC2000009950.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000426753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000426753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000428772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000428772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000439507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000439507.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
ENST00000457508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
NR_109916.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
NR_109917.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170403.hg.1 |
NR_109918.1 |
Refseq |