Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01570066_s1
          See other LOC107984208 GE Assays ›
          Gene Symbol:
          LOC107984208 LOC439951
          Gene Name:
          putative uncharacterized protein LOC439951
          uncharacterized LOC439951
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.10: 11610635 - 11612711 on Build GRCh38
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01570066_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC107984208
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017017017.1 XP_016872506.1 1 1327 59
          Gene Symbol: LOC439951
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC037281.1 - 1 666 4838780 59

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CELF2 antisense RNA 1 CELF2-AS1 C10orf31 -266.0kb Anti-sense Chr. 10 - 11316833 - 11344674 on NCBI Build GRCh38
          Downstream enoyl-CoA hydratase domain containing 3 ECHDC3 129.6kb Sense Chr. 10 - 11742357 - 11764070 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16291853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075364 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075389 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075382 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075385 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075396 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075400 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075376 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075409 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16291858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075380 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07075369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27430243 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P203953 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P203953 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 44912 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 24430 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P203953 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_017778_at - -
          Affymetrix Human Gene 1.0 ST Array XR_017773_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1555209_PM_at - -
          Affymetrix Human Genome U219 Array 11753389_at - -
          Affymetrix Human Genome U219 Array 11753389_PM_at - -
          Human Genome U133 Plus 2.0 Array 1555209_PM_at - -
          Human Genome U133 Plus 2.0 Array 1555209_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1555209_at4 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1694707 BC037281.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1694707 BC037281.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1694707 BC037281.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1694707 BC037281.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1694707 BC037281.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1694707 BC037281.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1694707 BC037281.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0