Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01572685_m1
          See other RIPK2 GE Assays ›
          Gene Symbol:
          RIPK2
          Gene Name:
          receptor interacting serine/threonine kinase 2
          Gene Aliases:
          CARD3, CARDIAK, CCK, GIG30, RICK, RIP2
          Chromosome Location:
          Chr.8: 89757747 - 89791064 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01572685_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RIPK2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003821.5 NP_003812.1 4 - 5 951 149
          XM_005251092.3 XP_005251149.1 3 - 4 761 149
          XM_011517357.2 XP_011515659.1 2 - 3 284 149
          GenBank mRNA AF027706.1 - 4 - 5 861 149
          AF064824.1 - 4 - 5 879 149
          AF078530.1 - 4 - 5 637 149
          AK075213.1 - 4 - 5 855 149
          AK301448.1 - 3 - 4 710 149
          AY358813.1 - 3 - 4 708 149
          AY358814.1 - 4 - 5 888 149
          AY524045.1 - 4 - 5 724 149
          BC004553.2 - 4 - 5 839 4026156 149
          KU178437.1 - 4 - 5 637 149
          KU178438.1 - 3 - 4 483 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800008179.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000545.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RIPK2

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine-mediated signaling pathway
          adaptive immune response
          apoptotic process
          inflammatory response
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          JNK cascade
          positive regulation of peptidyl-threonine phosphorylation
          peptidyl-tyrosine phosphorylation
          positive regulation of protein ubiquitination
          lipopolysaccharide-mediated signaling pathway
          positive regulation of chemokine production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          positive regulation of immature T cell proliferation
          positive regulation of peptidyl-serine phosphorylation
          toll-like receptor 2 signaling pathway
          toll-like receptor 4 signaling pathway
          T cell proliferation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          innate immune response
          positive regulation of T-helper 1 cell differentiation
          positive regulation of transcription by RNA polymerase II
          positive regulation of JNK cascade
          positive regulation of alpha-beta T cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of ERK1 and ERK2 cascade
          nucleotide-binding oligomerization domain containing signaling pathway
          nucleotide-binding oligomerization domain containing 1 signaling pathway
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          response to interleukin-1
          response to interleukin-12
          response to interleukin-18
          cellular response to lipoteichoic acid
          cellular response to peptidoglycan
          cellular response to muramyl dipeptide
          positive regulation of xenophagy
          protein serine/threonine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signaling receptor binding
          protein binding
          ATP binding
          LIM domain binding
          protein homodimerization activity
          CARD domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000518673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000518673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08007440.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08007440.hg.1 ENST00000540020.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08007440.hg.1 NM_003821.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08007440.hg.1 TC08000545.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0800156137.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800156137.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800156137.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800156137.hg.1 TC0800008179.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800156137.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay PSR0800156137.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0800156137.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000220751.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000517696.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000518673.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000518673.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 ENST00000522965.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq
          Affymetrix Human Clariom D Assay TC0800008179.hg.1 NM_003821.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0