Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017158.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017158.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017158.hg.1 |
TC09001375.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010913.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174256.hg.1 |
ENST00000375256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174256.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174256.hg.1 |
TC0900010913.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900174256.hg.1 |
NM_153695.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174256.hg.1 |
ENST00000375256.4 |
Ensembl |