Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01573471_m1
          See other PTGS2 GE Assays ›
          Gene Symbol:
          PTGS2
          Gene Name:
          prostaglandin-endoperoxide synthase 2
          Gene Aliases:
          COX-2, COX2, GRIPGHS, PGG/HS, PGHS-2, PHS-2, hCox-2
          Chromosome Location:
          Chr.1: 186671812 - 186680427 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01573471_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTGS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000963.3 NP_000954.1 3 - 4 455 111
          GenBank mRNA AJ634912.1 - 3 - 4 318 111
          AK292167.1 - 3 - 4 443 111
          AY151286.1 - 3 - 4 415 111
          AY462100.1 - 3 - 4 318 111
          BC013734.1 - 3 - 4 438 3880850 111
          DA277667.1 - 3 - 4 111
          L15326.1 - 3 - 4 410 111
          M90100.1 - 3 - 4 415 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100016715.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003638.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PTGS2

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          angiogenesis
          prostaglandin metabolic process
          nicotinamide metabolic process
          movement of cell or subcellular component
          inflammatory response
          response to oxidative stress
          embryo implantation
          learning
          memory
          regulation of blood pressure
          negative regulation of cell proliferation
          response to fructose
          response to manganese ion
          response to lithium ion
          positive regulation of vascular endothelial growth factor production
          sensory perception of pain
          cyclooxygenase pathway
          lipoxygenase pathway
          bone mineralization
          ovulation
          positive regulation of prostaglandin biosynthetic process
          positive regulation of fever generation
          positive regulation of synaptic plasticity
          negative regulation of synaptic transmission, dopaminergic
          response to estradiol
          response to lipopolysaccharide
          response to vitamin D
          response to tumor necrosis factor
          cellular response to UV
          maintenance of blood-brain barrier
          positive regulation of NF-kappaB import into nucleus
          response to drug
          hair cycle
          positive regulation of apoptotic process
          positive regulation of nitric oxide biosynthetic process
          negative regulation of cell cycle
          positive regulation of vasoconstriction
          negative regulation of smooth muscle contraction
          positive regulation of smooth muscle contraction
          decidualization
          positive regulation of smooth muscle cell proliferation
          regulation of inflammatory response
          brown fat cell differentiation
          response to glucocorticoid
          negative regulation of calcium ion transport
          positive regulation of synaptic transmission, glutamatergic
          oxidation-reduction process
          response to fatty acid
          cellular response to mechanical stimulus
          cellular response to ATP
          cellular response to hypoxia
          cellular response to fluid shear stress
          positive regulation of transforming growth factor beta production
          positive regulation of cell migration involved in sprouting angiogenesis
          positive regulation of fibroblast growth factor production
          positive regulation of brown fat cell differentiation
          positive regulation of platelet-derived growth factor production
          cellular oxidant detoxification
          peroxidase activity
          prostaglandin-endoperoxide synthase activity
          protein binding
          lipid binding
          enzyme binding
          heme binding
          protein homodimerization activity
          metal ion binding
          arachidonate 15-lipoxygenase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01055966.hg.1 TC01003638.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01055966.hg.1 ENST00000367468.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055966.hg.1 ENST00000490885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055966.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000466691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000466691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000466691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100226827.hg.1 TC0100016715.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000367468.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000466691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000466691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000466691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000490885.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559627.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 ENST00000559800.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq
          Affymetrix Human Clariom D Assay TC0100016715.hg.1 NM_000963.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0